missing translation for 'onlineSavingsMsg'
Learn More

Brix Standard, Reagecon™
BIOPHARMA

Product Code. 13494036 Shop All Reagecon Products
Change view
Click to view available options
Concentration:
10% sucrose
30% sucrose
5% sucrose
60% sucrose
Unit Size:
15mL
Product Code. Concentration unitSize
13494036 60% sucrose 15mL
13474016 10% sucrose 15mL
13474026 30% sucrose 15mL
13494016 5% sucrose 15mL
4 options
This item is not returnable. View return policy

Product Code. 13494036

Brand: Reagecon BS60S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Stabilized to have an extended 12-month shelf-life. Reagecon™ Brix Standard is a range of ready-to-use sucrose refractive index standards in a convenient dropper bottle for use with all types of refractometers. Test results accredited to ISO 17025 and manufactured in accordance with ISO Guide 34.

This range of sucrose standard solutions contains high purity raw ingredients, can be used with any brand of refractometer and is stabilized to have an extended shelf-life to provide excellent value for users that are not required to follow ICUMSA Guidelines.

  • Prepared gravimetrically on a weight/weight basis: both solute (sucrose) and solvent (water) are weighed on a balance calibrated using OIML traceable weights
  • Balance Certificate of Calibration is issued in accordance with ISO 17025 for calibration of nonautomatic weighing machines (INAB ref: 265C)
  • Brix value is verified using a high-performance calibrated, temperature-controlled refractometer using high-purity, ISO Guide 34-accredited Brix standards

Chemical Identifiers

CAS 57-50-1 , 7732-18-5
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00006626
InChI Key CZMRCDWAGMRECN-PWPRYFECNA-N
Synonym Sucrose solution
PubChem CID 5988
ChEBI CHEBI:17992
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O

Specifications

Description Accredited to ISO 17025, In accordance with ISO Guide 34
Packaging Dropper bottle
Physical Form Liquid
Concentration 60% sucrose
For Use With (Equipment) Refractometer
Solubility Soluble in water
IUPAC Name (2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Solvent or Matrix Water
Quantity 15 mL
Formula Weight 342.3g/mol
Chemical Name or Material Sucrose
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.