Learn More
VACUUBRAND™ Service Kits for NT Series Chemistry Pumps and Systems
Product Code. 15960877 Shop All Vacuubrand Products
Change view
Click to view available options
For Use With:
MD 4C NT VARIO, PC3003/4 VARIO
ME 4C NT
ME 8C NT
MZ 2C NT/VARIO, PC201 NT, PC3002 VARIO
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With unitSize
15960877 MZ 2C NT/VARIO, PC201 NT, PC3002 VARIO Each
16254312 ME 4C NT Each
10404813 ME 8C NT Each
10355303 MD 4C NT VARIO, PC3003/4 VARIO Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15960877 Supplier VACUUBRAND™ Supplier No. 20696869
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Service kits for VACUUBRAND NT series Chemistry design pumps.
  • Sets include appropriate diaphragms, valves and sealing rings (if applicable)
  • Wrench not included.
TRUSTED_SUSTAINABILITY
Specifications
For Use With MZ 2C NT/VARIO, PC201 NT, PC3002 VARIO
Type Service Kit