missing translation for 'onlineSavingsMsg'
Learn More

Velp Scientifica™ BOD Sensor

Product Code. 11795894 Shop All Velp Products
Click to view available options
Battery Type:
2 x 3 V Lithium
Unit Size:
Each
This item is not returnable. View return policy
This item is not returnable. View return policy

Velp Scientifica™ BOD Sensor is innovative, mercury-free and extremely reliable solution for BOD analysis (Biochemical Oxygen Demand).

Precise Control

  • Measurement on scales of 90, 250, 600 and 999 ppm BOD
  • Higher values can be measured by diluting the sample
  • Models with stirring stations offer VELP stirring quality


Extremely Practical

  • Fits directly on the bottle containing the sampl
  • Automatically stores 5 BOD values at 24-hour intervals
  • Enables analysis to continue over the weekend
  • Results directly readable at any time, even after five days
  • Direct readout in mg/l (ppm)
  • Compact stirring stations

Specifications

Battery Type 2 x 3 V Lithium
For Use With (Equipment) BOD Analysis
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.