Learn More
IBA Lifesciences BioLock
GREENER_CHOICE
Product Code. 15643608 Shop All IBA Lifesciences Products
missing translation for 'orderingAttributeHoverText'
Capacity (Metric):
50 mL
250 mL
missing translation for 'unitSize'
250mL
50mL
This item is not returnable. View return policy
This item is not returnable. View return policy
BioLock is a Biotin blocking solution for lysates and culture supernatants
BioLock is a ready-to-use biotin blocking solution which contains the tetrameric biotin binding protein avidin. BioLock prevents biotin and biotinylated proteins from binding to Strep-Tactin™ or Strep-Tactin™XT, whereas binding of the Twin-Strep-tag™ and Strep-tag™II is not influenced.
Biotin and/or biotinylated proteins can possibly reduce the binding capacity of Strep-Tactin™ as well as Strep-Tactin™XT and are present in cell lysates and cell culture media, especially from mammalian or insect cells. Thus, if Twin-Strep-tag™ and Strep-tag™II fusion proteins from biotin-containing extracts or media are intended to be purified via Strep-Tactin™ or Strep-Tactin™XT affinity chromatography, biotin has to be masked by addition of BioLock prior to application onto the resin. 
Specifications
Capacity (Metric) 50 mL
Type Blocking Solution
For Research Use Only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.