Learn More
Novacyt - Microgen Bioproducts Biochemical Identification System: Staph spp.
Product Code. 15997112 Shop All Novacyt Group Products
Change view
Click to view available options
Quantity:
20 Tests
Unit Size:
20 tests
Product Code. Quantity unitSize
15997112 20 Tests 20 tests
1 options
This item is not returnable. View return policy
Product Code. 15997112
Brand: Novacyt - Microgen Bioproducts MID69CE
Please to purchase this item. Need a web account? Register with us today!
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
This item is not returnable. View return policy
Biochemical identification system for Staph spp. from foods and other environmental samples.
  • Twelve-substrate biochemical identification system.
  • From a range of different technologies for the specific detection of Staph spp. from foods and other environmental samples.
  • From a range of simple, reliable, bacterial identification systems for food, environmental, and industrial laboratories.
  • These tests will arm professionals with the necessary information to maximize the effectiveness of appropriate patient management.
  • The range is expanding quickly and will encompass the widest range of microbiological testing solutions.
Specifications
For Use With (Application) Identification of Staph Spp.
Quantity 20 Tests