missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek Synergy LX Multi-Mode Reader
GREENER_CHOICE

The BioTek Synergy LX multi-mode reader automates many common microplate assays. NOTE: Fluorescence filter cubes are required for fluorescence measurements (sold separately). For Research Use Only. Not for use in diagnostic procedures.

Brand:  Agilent BioTek SLXATS-SN

 View more versions of this product

Product Code. 17267829

  • 110850.00 DKK / Each

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The BioTek Synergy LX multi-mode reader automates many common microplate assays. The high-quality optic design ensures excellent data in absorbance, fluorescence, and luminescence detection modes. This affordable system is ideal for many common assays including nucleic acid and protein quantification, ELISA, BCA, Bradford, and cell viability.

The data display, output to a USB flash drive, printer, or Gen5 software make Synergy LX a versatile assay workstation. The large touch screen user interface and onboard software simplify programming and save time in operation of microplate and microvolume assays.

NOTE: Fluorescence filter cubes are required for fluorescence measurements (sold separately).

Certifications

CE and TUV marked. RoHS compliant.

Services

12 Months

TRUSTED_SUSTAINABILITY
Specifications

Specifications

200 to 900 nm
CE and TUV marked. RoHS compliant.
38.1 cm
24 sec. (96 wells), 76 sec. (384 wells)
Monochromator
Nucleic acid and protein quantification, ELISA, BCA, Bradford and cell viability
50/60 Hz
<1% from 0 to 2.5 OD
Absorbance
Multi-mode Microplate Reader
0.001 OD
2
1 year
27 lb.
15 in.
Photodiode, PMT
End Point, Kinetic, Area Scanning, Absorbance Spectral Scanning
Wavelength: ±2 nm; Optical Density: <1% at 2.0 OD, <3% at 2.5 OD
≤5 nm
15 in.
Linear, Orbital, Double Orbital
Monochromator Bandpass
15 in.
38.1 cm
Reader with Touchscreen
SLXATS
24 VDC power supply compatible with 100/240 VAC at 50/60 Hz, 60 W (max.)
Fluorescence Intensity: Fluorescein 2 pM; Luminesence: 10 amol ATP
English
100/240 V
12.3 kg
38.1 cm
Xenon Flash
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.