Learn More
Applied Biosystems™ BigDye™ Terminator v3.1 Cycle Sequencing Kit
The BigDye™ Terminator v3.1 Cycle Sequencing Kit's robust, highly flexible chemistry is ideal for de novo sequencing, resequencing, and finishing with PCR product, plasmid, fosmid, and BAC templates.
Brand:  Applied Biosystems™ 4337458
264005.00 EUR valid until 2025-12-31
Use promo code "24365" to get your promotional price.
 View more versions of this product
Product Code. 17152979
  • 291500.00 € / 25000 reactions
Please to purchase this item. Need a web account? Register with us today!
For Research Use Only. All usage must comply with product instructions.
This item is not returnable. View return policy
Description
Description
The BigDye™ Terminator v3.1 Cycle Sequencing Kit's robust, highly flexible chemistry is ideal for de novo sequencing, resequencing, and finishing with PCR product, plasmid, fosmid, and BAC templates.
  • Improve the quality of your results for a wide range of sequencing applications
  • Optimized for long read lengths
  • Better dye mobility characteristics
  • Improved performance reading through GT-rich regions
  • Get longer, higher-quality reads with more uniform peak heights and optimal signal balance
  • Enhance your productivity and reduce costs
Get more uniform peak heights and improved accuracy
With BigDye™ Terminator v3.1 kit superior chemistry, you generate data with uniform peak heights and optimized signal balance. This results in longer, higher-quality reads and more accurate base assignments for heterozygote and mutation detection.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
• 10 x 20 mL tubes of BigDye™ Terminator v3.1 Ready Reaction Mix
• 4 tubes M13 (-21) Primer
• 4 tubes pREF-BDT™ Control DNA
• 1 x 233 mL tube of 5X Sequencing Buffer (may be stored at 4°C)
Store kit at -15°C to -25°C, and avoid repeated freeze-thaw cycles.
SeqStudio™ Genetic Analyzer, SeqStudio™ Flex Series Genetic Analyzer, 3500/3500xL Genetic Analyzer, 3730/3730xl DNA Analyzer Thermal Cyclers: GeneAmp™ PCR System 9700 Dual Series, Veriti™ Fast 96‐well and 384‐well Thermal Cyclers
BigDye™ Terminator
25,000 Reactions
BAC DNA, Plasmid DNA (≤15Kb), PCR Amplicons, Single Stranded DNA
12-well Strips, 8-well Strip, 96- or 384-well Plate
Standard Sequencing
Sequencing Buffer
Fluorescent Dye Terminator Sequencing
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
For Research Use Only. Not for use in diagnostic procedures.