missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Sterileware™ V-shaped Sampling Spatula, Sterile

Product Code. 11897822 Shop All SP Scienceware Products
Click to view available options
Length (Metric):
22.9 cm
Unit Size:
Pack of 100
This item is not returnable. View return policy
This item is not returnable. View return policy

Ideal for every type of powdered or granulated materials. Spatulas are ideal for taking sterile samples during process quality inspections and for piercing plastic-lined paper bags or similar packages.

  • Time-honored V-shape makes transferring powdered, flaked, crystalline or granular media a breeze.
  • Long, 14 in. spatula (16-241-210) is perfect for dislodging caked or crystallized reagents from deep within tall bottles or vessels having a neck diameter as small as 0.75 in. (18 mm).
  • Sterileware (16-241-210, 14-385-20) available in 9 and 14 in. models.
  • High-impact polystyrene.
  • Individually packaged.
  • Sterilized by gamma irradiation.
  • Lot stamped, shipped with Certificate of Processing.
  • Nonsterile Model (03-421-149) 9 in. spatula.

Specifications

Sterility Sterile
Autoclavable Not Autoclavable
Shape Pointed End
Disposable Single-use
Product Type Sterileware™ Sampling Spatula
For Use With (Equipment) Containers, bags and drums
For Use With (Application) Ideal for every type of powdered or granulated material, perfect for taking sterile samples during process quality inspections and for piercing Plastic-lined Paper bags or similar package
Length (English) 9 in.
Width (English) 0.75 in.
Width (Metric) 1.9 cm
Dimensions (L x W) 9 x 0.75 in. (22.9 x 1.9 cm)
Includes 6 in. L V-shaped channel
Length (English) Handle 2.99 in.
Length (Metric) Handle 7.6 cm
Material Polystyrene
Length (Metric) 22.9 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.