Learn More
Bel-Art™ Secador™ Mini Desiccator Cabinet
Assures easy and reliable storage of moisture sensitive products in a space saving size. Bel-Art™ SP Scienceware™ Secador™ Mini Desiccator Cabinet is a great choice when benchtop space is at a premium.
Brand:  Bel-Art™ F42075-1000
Product Code. 11862742
  • 196.00 € / Each
Spotlight
Use promo code 24158 at checkout
View Available Promotions
Only null left
In Stock
Edge
Add to basket
Description
Description
Assures easy and reliable storage of moisture sensitive products in a space saving size. Bel Art™ SP Scienceware™ Secador™ Mini Desiccator Cabinet is a great choice when benchtop space is at a premium.
  • Large side swinging door maximizes access to interior space, has a patented seal design, tab latches and security loops for padlock or tamper-evident seals, opens right to left.
  • Easy-to-view dial hygrometer located in the door front.
  • Clear polystyrene for easy viewing of contents.
  • Can be used with desiccant cartridges F42048-0065, F42048-0100 or F42046-0000.
  • Kit available to mount to the underside of a shelf (F42075-0201).
  • Overall Dimensions: 33.8 W x 25.4 D x 21.6 cm H (13.3 x 10 x 8.5 in.).
  • Internal Volume: 0.3 cu. ft.
  • Polystyrene does not block UV light: for UV protection, select a Secador 1.0 model or larger.
Specifications
Specifications
Clear
Copolyester, Polystyrene
Latched, swinging
2 lockable loops with padlock or tamper-evident seals, door-mounted dial hygrometer
13.3 in.
33.8 cm
8.5 in.
21.6 cm
25.4 cm
Front-loading
Desiccator
For small moisture-sensitive objects and substances
0.35 cu. ft.
9.3 in.
23.6 cm
5 in.
10 in.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.