missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Polypropylene Coplin Staining Jar

Product Code. 10082844 Shop All SP Scienceware Products
Click to view available options
Slide Count:
10
Unit Size:
Each
This item is not returnable. View return policy
This item is not returnable. View return policy

Improves on the classic tapered square design with a larger opening for easier placement and removal of slides. Bel Art™ Polypropylene Coplin Staining Jar makes working with slides easier.

 

    • Five internal slots hold up to 10 standard 25 x 75mm slides back to back
    • Steam autoclavable at 121ºC
    • Flat top
    • Easy grip knurled edge
    • Deep thread
    • Seal ring to prevent leaks

 

Specifications

Autoclavable Autoclavable
Bottom Shape Round
Capacity (Metric) 322 cm3
Color White
Disposable Reusable
Material Polypropylene
Shape Cylindrical
Sterility Non-sterile
Surface Treatment Non-treated
Type Staining Jar
Quantity Each
Slide Count 10
Height (Metric) 11.4 cm
Height (English) 4.5 in.
Diameter (English) 2.375 in.
Diameter (Metric) 59.7 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.