missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Flowmi™ Cell Strainers for 1000μL Pipette Tips
Provides fast and efficient filtering of small volume samples (up to 1000 μL) prior to FLOW or FACS analysis (patent pending). Available in two porosity sizes.
Brand: Bel-Art™ H13680-0070
Description
- Provide fast and efficient filtering of small volume samples (up to 1000 μL) prior to FLOW or FACS analysis
- Preserves volume when filtering small sample volumes
- Removes cell aggregates and creates uniform single cell suspensions
- Decreases the potential for clogging of FLOW or FACS instruments by effectively filtering cellular debris
- Compact tray holds 50 Flowmi Cell Strainers ready for press and fit attachment to tip; features a sliding cover for easy, one-handed use
- Packaged in a re-sealable bag to maintain sterility when used in laminar flow hood or biohazard hood
- Sterile, 40 μm or 70 μm porosity
Specifications
Polyethylene | |
Provide fast and efficient filtering of small volume samples (up to 1000μL) prior to FLOW or FACS™ analysis | |
Cell Strainer | |
Pipette Tips, 1000 μL, including Fisherbrand Sure-One, VWR, Axygen, Nichiryo, Eppendorf and more |
White | |
Compact tray | |
Sterile | |
70 μm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction