missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Clavies™ Heat-Resistant Biohazard Autoclave Gloves

Soft, pliant, cotton terrycloth gloves are ideal for handling hot objects out of the autoclave or oven and protect from temperatures up to 450°F (232°C).

Brand:  Bel-Art™ H13201-0001

391.27 DKK valid until 2025-06-30
Use promo code "27322" to get your promotional price.


 View more versions of this product

Product Code. 10762682

  • 496.00 DKK / 1 pair
Spotlight
Use promo code 24156 at checkout
View Available Promotions
Only null left
Estimated Shipment: 18-07-2025
Add to basket
This item is not returnable. View return policy

Description

Description

Soft, pliant, cotton terrycloth gloves are ideal for handling hot objects out of the autoclave or oven and protect from temperatures up to 450°F (232°C).

  • Ideal for removing hot objects from an autoclave or oven
  • Heat-resistant up to 232°C
  • Replaces hazardous asbestos gloves
  • Extended gauntlet cuff available for additional forearm protection
  • Gloves are not waterproof and should not be used to handle dry ice
  • Machine wash, air dry, or use the Lab-Aire™ II Glove Holder (F18931-0000) to dry on any Lab-Aire™ dryer
Specifications

Specifications

CE
Laboratory, Industrial
Orange
Terrycloth
Universal
450°F
Handling Hot Objects
H13201
11 in.
28 cm
Yes
Uncoated
Yes
Autoclave Gloves
Full-hand Mitt
232°C
Gauntlet
18.5 in.
47 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.