missing translation for 'onlineSavingsMsg'
Learn More

Barium Sulfate, U.S.P., J.T. Baker™

Product Code. 15577884 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15577884 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15577884 Supplier Avantor J.T.Baker Supplier No. 104001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 7727-43-7
Quantity 500 g
Molecular Formula BaO4S
MDL Number MFCD00003455
Synonym barium sulfate, barite, baritop, barosperse, barotrast, esophotrast, liquipake, macropaque, micropaque, microtrast
InChI Key TZCXTZWJZNENPQ-UHFFFAOYSA-L
SMILES [Ba++].[O-]S([O-])(=O)=O
IUPAC Name barium(2+);sulfate
Molecular Weight (g/mol) 233.38
PubChem CID 24414
Formula Weight 233.39
Grade USP
Packaging Poly Bottle
Specific Gravity 1L = 4.50kg
Chemical Name or Material Barium Sulfate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.