missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Aerosol Barrier Micropipette Tips

Product Code. 11366054 Shop All Axygen Scientific Products
Change view
Click to view available options
Volume (Metric):
0.5 to 50 μL
1 to 150 μL
100 to 1000 μL
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 1000
Pack of 10000
Pack of 960
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Sterility unitSize
11366054 1 to 150 μL Sterile Pack of 960
12541358 0.5 to 50 μL Non-sterile Pack of 10000
12605475 0.5 to 50 μL Non-sterile Pack of 1000
12665475 100 to 1000 μL Non-sterile Pack of 1000
12581348 100 to 1000 μL Non-sterile Pack of 10000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11366054 Supplier Axygen™ Supplier No. TF150RS
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Axygen™ Aerosol Barrier Micropipette Tips allow free passage of air while blocking aerosol contamination.

  • Technologically advanced filter material
  • Filter membrane contains no leachable additives
  • Available with Maxymum Recovery™ surface for improved accuracy and maximum sample recovery
  • Available in bulk, or racked (sterile)
  • Available in a variety of material contact tip options
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1 to 150 μL
Filtered Filtered
Sterility Sterile
Color Clear
Autoclavable Autoclavable
Clean Claims DNase, RNase Free
Format Rack
Tip Style Universal
Packaging 96 tips/rack, 10 racks/pack, 5 packs/case
Product Type Aerosol Barrier Micropipette Tips
For Use With (Equipment) Universal Fit
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.