Learn More
Thermo Scientific™ Automation Tips for Caliper™ Zymark™ Liquid Handling Workstations
Shop All MBP Molecular BioProducts Products

Description
Increase pipetting performance with Thermo Scientific™ Automation Tips for Caliper™ Zymark™ Liquid Handling Workstations, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Caliper/Zymark RapidPlate, Caliper Presto, Allegro, Zephyr and SciClone.
Features:
Specifications
Specifications
| Volume (Metric) | 100 μL |
| Filtered | Non-filtered |
| Sterility | Non-sterile |
| Color | Clear |
| Format | Rack |
| Tip Style | Automation |
| For Use With (Equipment) | Zymark/Caliper Liquid Handling Workstations |
| Product Line | Caliper Zymark |
| Fit for Purpose | Certified Quality |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.