missing translation for 'onlineSavingsMsg'
Learn More

3M™ Aura™ Particulate Respirators 9300+ Series

Product Code. p-8001313
Click to view available options
Filter Type:
FFP1
FFP2
FFP3
With Exhalation Valve:
No
Yes
Assigned Protection Factor (APF):
4
10
20
Model:
9310
9312
9320
9322+
9322+ P2 valved
9330
9330+ P3
9332
9332+ P3 valved
PF9312
PF9320
PF9322
PF9332
Unit Size:
Each
Pack of 10
Pack of 120
Pack of 20
Pack of 240
Pack of 330
Pack of 440
Pack of 5
This item is not returnable. View return policy
This item is not returnable. View return policy

Particulate respirators have an ergonomic, flat-fold, 3-panel design featuring a low breathing resistance filter media. Available in FFP1, FFP2, and FFP3 protection levels with or without valves, Innovative chin tab for easy adjustments and an embossed, sculpted upper panel helps reduce eyewear fogging.

  • Provide comfortable protection against dusts and mists in a wide variety of industrial applications and other hazardous environments.
  • Offers FFP1, FFP2 and FFP3 protection.
  • 3-panel design allows for greater facial movement.
  • Sculpted, low-profile style of the embossed upper panel creates an effective seal, reduces eyewear fogging and improves eyewear compatibility.
  • Low resistance filter technology promotes easier breathing and the innovative chin tab allows simple adjustments.
  • Choose without valve style or with valve styles that feature 3M™ Cool Flow™ to reduce heat build-up and help provide comfortable protection in hot and humid conditions.
  • Individually packed in hygienic wrapping to prevent contamination before use.
  • Braided, colour-coded headbands help you identify the protection level offered: yellow for FFP1 (APF 4), blue for FFP2 (APF 10) and red for FFP3 (APF 20).
  • Foldable design makes them easy to store when not in use.
  • Respirators work well with 3M eyewear and hearing protection products.
TRUSTED_SUSTAINABILITY

Specifications

Assigned Protection Factor (APF) 4
Certifications/Compliance CE Approved to EN 149:2001A1:2009
Certifications/Compliance (Europe) EN 149:2001 + A1:2009
Certifications (EN) EN 149:2001 + A1:2009
Color White
Design Type Flat-fold
Disposable Single-use
Filter Type FFP1
Material Polypropylene
Product Type General Purpose Particulate Respirator
Size One Size
With Exhalation Valve No
Filtration Layer Material Polypropylene
For Use With (Application) Agriculture, Cleaning Maintenance, Demolition
Latex Free Yes
Material Headstrap Polyisoprene
Model 9310
Nose Piece Material Aluminum, Polyurethene
Nosepiece Type Sculpted
Packaging Type Individually Packaged
Quantity 20/Pk.
Resistant To Dusts and mists found in a wide variety of industrial applications and other work situations
Shape Cup
Sterility Non-sterile
Style 3-panel
Strap Attachment Type Colour-coded Headband, Yellow
FDA Cleared Surgical Mask No
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.