missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arsenic Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
60.68 EUR - 135.00 EUR
Chemical Identifiers
CAS | 7440-38-2, 7697-37-2, 7732-18-5 |
---|---|
Molecular Formula | As |
Molecular Weight (g/mol) | 74.92 |
MDL Number | MFCD00085309 |
InChI Key | RQNWIZPPADIBDY-UHFFFAOYSA-N |
PubChem CID | 5359596 |
ChEBI | CHEBI:27563 |
IUPAC Name | arsenic |
SMILES | [As] |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10171762
|
SPEX Certiprep
PLAS2-2Y |
125 mL | Triple-leached LDPE bottle |
60.68 EUR
125mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11468619
|
SPEX Certiprep
PLAS2-2T |
250 mL | Triple-leached LDPE bottle |
98.70 EUR
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10224242
|
SPEX Certiprep
PLAS2-2X |
500 mL | Triple-leached LDPE bottle |
135.00 EUR
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
As | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
7440-38-2, 7697-37-2, 7732-18-5 | |
74.92 | |
RQNWIZPPADIBDY-UHFFFAOYSA-N | |
CHEBI:27563 | |
[As] |
As | |
MFCD00085309 | |
5359596 | |
arsenic |
Specifications
7440-38-2, 7697-37-2, 7732-18-5 | |
As | |
AA, ICP | |
RQNWIZPPADIBDY-UHFFFAOYSA-N | |
[As] | |
2% HNO3 | |
5359596 | |
Assurance |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
MFCD00085309 | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
Certified Reference Material | |
arsenic | |
74.92 | |
CHEBI:27563 | |
Arsenic Standard |
Safety and Handling
Causes severe skin burns and eye damage.
missing translation for 'dotInformation' : Hazardous
Recommended Storage : Ambient
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 1000
Signal Word
- Danger||Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : nitric acid, arsenic
Product intended for laboratory use only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title