missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 02-100 Chemical Resistant, 5 Layer Laminated Gloves

Product Code. 12313609 Shop All Ansell Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12313609 7 12 pairs
12689781 6 12 pairs
11517853 8 12 pairs
12333609 9 12 pairs
12343609 10 12 pairs
15399622 11 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 12313609 Supplier Ansell Supplier No. 2100070
Only null left
Add to basket
Edge
Add to basket

Laminated gloves provide extreme resistance against a wide range of chemicals, including biological hazards.

  • Provide flexibility and tactility
  • Made from an exceptionally thin chemical-resistant, five-layer laminated film
  • Integral, nonwoven liner helps absorb and dissipate perspiration for a cooler, more comfortable fit
  • Can be worn as liners under heavier nitrile, neoprene, or PVC gloves
  • Hand specific
  • Broad spectrum chemical resistance
  • Wing-thumb for added wearer comfort
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) CE 0493, EN420:2003 + A1:2009; Category lll
Applications Industrial
Color White
Latex Free Yes
Material Laminated Film
Product Type Chemical-resistant Gloves
Size 7
Thickness (English) 2.5 mil
Thickness (Metric) 0.06 mm
EN388 Standard EN 388:0000
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES ABCEFGHIMS
EN ISO 374 - 5 Bacteria, Fungi WITH Virus Protection
Grip Design Smooth
Lining Unlined
Model 02-100_7
Series AlphaTec™ 02-100
Length (English) 15 in.
Length (Metric) 38 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.