missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Formate in Methanol, 10mM, with 0.05% Formic Acid, LC-MS

Product Code. 15203075 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 L
Packaging:
Borosilicate Glass Bottle
Unit Size:
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Mobile phase blend suitable for analysis such as vitamin D and drugs of abuse testing, and packaged in a borosilicate glass bottle that can be used directly on the LC-MS instrument. | CAS: 540-69-2 | CH4O | 32.04 g/mol

Ammonium Formate in Methanol, 10mM, with 0.05% Formic Acid, 0.57 to 0.69g/L (Ammonium Formate, w/v), CAS Number-540-69-2, 67-56-1, 64-18-6

  • Suitable for use in LC/MS and UHPLC applications
  • Pre-mixed for ease-of-use
  • Mobile phase blend suitable for vitamin D and drugs of abuse testing  
  • Packaged in borosilicate glass bottles to prevent metal adduct formation, which can be used directly on the instrument
  • Ammonium formate- a commonly used HPLC buffer that is compatible with LC/MS
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 64.7°C
Chemical Name or Material Ammonium Formate in Methanol, 10mM, with 0.05% Formic Acid
Name Note Mobile-phase solvent blend
CAS 540-69-2, 67-56-1, 64-18-6
pH 3.5 (25°C)
Physical Form Liquid
Quantity 1 L
Molecular Formula CH4O
MDL Number MFCD00004595
Synonym ammonium formate, formic acid, ammonium salt, formic acid ammonium salt, ammoniumformate, azanium formate, ammonium formiate, formic acid, ammonium salt 1:1, mravencan amonny czech, hsdb 479, mravencan amonny
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) 32.04
Grade UHPLC-MS
Concentration or Composition (by Analyte or Components) 10mM Ammonium Formate
Packaging Borosilicate Glass Bottle
Vapor Pressure 128 at 20°C
Density 0.791 g/cm3
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 10mM Ammonium Formate in Methanol + 0.05% Formic Acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
  • Flammable liquid Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
Supplemental information
  • MIXTURE LIST-Contains : Methyl alcohol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.