missing translation for 'onlineSavingsMsg'
Learn More

Ammonium fluoride - hydrofluoric acid mixture, Honeywell™

Product Code. 15713958 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
2.5 L
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15713958 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15713958 Supplier Honeywell Research Chemicals Supplier No. 40207H2.5L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Semiconductor Grade, AF 875-125, etching mixture (Honeywell 17621)

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1341-49-7, 12125-01-8, 7732-18-5
Boiling Point 100°C
CAS Min % ≥10.00%, ≥20.00%, ≥50.00%
CAS Max % <20.00%, <30.00%, <70.00%
Purity Grade Notes Semiconductor grade reagent
Packaging Plastic Bottle
Quantity 2.5 L
Physical Form Liquid, clear
compliance-icons compliance-icons
Product Identifier
  • Ammonium fluoride - hydrofluoric acid mixture
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : ammonium fluoride, ammonium bifluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.