missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Acetate, For HPLC, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 50 g |
| Linear Formula | CH3CO2NH4 |
| Beilstein | 4186741 |
| Solubility Information | Soluble in water 0.1g/mL at 25°C |
| Formula Weight | 77.08g/mol |
| Percent Purity | ≥99% |
| Chemical Name or Material | Ammonium Acetate |
Sikkerhed og håndtering
Produktidentifikator
- Ammonium acetate
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion