Learn More
Cytiva Amersham™ ECL Select™ Western Blotting Detection Reagent
Use as a high sensitivity reagent for chemiluminescent Western blotting detection. Cytiva Life Sciences™ Amersham™ ECL Select™ Western Blotting Detection Reagent provides highly intense signal output, for detection of medium to very low protein levels.
Brand:  Cytiva RPN2235
Product Code. 12644055
  • 414.00 € / Each
Please to purchase this item. Need a web account? Register with us today!
Edge
This item is not returnable. View return policy
Description
Description
Amersham ECL Select is a high sensitivity reagent for chemiluminescent Western blotting detection. High signal intensity makes it suitable for the most demanding Western blotting applications, allowing for detection of minute protein quantities. Signals can be detected with CCD imager or X-ray film. Amersham ECL Select typically exhibits a dynamic range that is complementary to Amersham ECL Prime™, and similar to Amersham ECL Advance™.
  • Amersham ECL Select is the most sensitive chemiluminescent Western blotting detection reagent in the Amersham ECL range
  • With exceptional signal intensity, Amersham ECL Select results in brighter, clearer bands at low protein levels
  • Amersham ECL Select delivers high sensitivity even when using highly diluted primary and secondary antibodies
  • Works with a wide range of antibody dilutions, reducing the need for lengthy optimization
  • Optimized for imaging with ImageQuant™ LAS 4000 (CCD-based imaging) and compatible with Amersham Hyperfilm
  • Compatible with Rainbow Molecular Weight Markers and Amersham ECL DualVue™ Western Blotting Markers
  • Optimized for use with Amersham Hybond™ PVDF membranes and compatible with Amersham Protran™ nitrocellulose membranes
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Chemiluminescence
High Signal Intensity Western Blot
Solution A (Luminol solution, 50mL), Solution B (Peroxide solution, 50mL)
Liquid
Nitrocellulose, PVDF
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • ECL Select
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360D-May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains :
  • UNKNOWN ACUTE-per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains % of components with unknown hazards to the aquatic environment.
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.