missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ ECL™ Prime Western Blotting Detection Reagent

Enables the use of highly diluted primary and secondary antibodies without reducing sensitivity. Cytiva Lifescience™ Amersham™ ECL™ Prime Western Blotting Detection Reagent produces high signal intensity and sensitivity.

Brand:  Cytiva RPN2236


 View more versions of this product

Product Code. 12994780

  • 704.00 EUR / Pack of 3

Please to purchase this item. Need a web account? Register with us today!

Edge

Includes: Solution A (luminol solution) and Solution B (peroxide solution)
This item is not returnable. View return policy

Description

Description

p>
  • Stable signal enables multiple exposures and makes reagent suitable for large experimental series
  • Provides convenient handling time between detection and completion of experiment
  • Optimized for imaging with ImageQuant™ LAS 4000 (CCD-based imaging), Amersham Hybond™ P (PVDF) membranes and compatible with Amersham Hyperfilm™, Amersham Hybond- ECL (nitrocellulose) membranes
  • Also compatible with Rainbow™ Molecular Weight Markers and Amersham ECL DualVue™ Western Blotting Markers

Education and research

TRUSTED_SUSTAINABILITY
Specifications

Specifications

ECL Western Blot Detection Reagent
∼1 pg
Solution A (Luminol solution, 3 × 50mL) and Solution B (Peroxide solution, 3 × 50mL)
Sensitive Quantitative or Confirmatory Western Blot
3000cm2 membrane
2°C to 8°C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • ECL Prime

Signal Word
  • Danger

Hazard Category
  • Reproductive toxicity Category 1B

Hazard Statement
  • H360D-May damage the unborn child.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : trometamol, ethanediol, imidazole
  • UNKNOWN AQUATIC-Contains 4% of components with unknown hazards to the aquatic environment.

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.