missing translation for 'onlineSavingsMsg'
Learn More

PCRmax Alpha Cycler 4 Thermal Cycler

Product Code. 15569164
Change view
Click to view available options
Capacity:
Dual 384-well, Dual 96-well Capacity
Quad 384-well Capacity
Single 384-well, Triple 96-Well Capacity
Triple 384-well, Single 96-Well Capacity
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
15569164 Single 384-well, Triple 96-Well Capacity Each
15522895 Dual 384-well, Dual 96-well Capacity Each
15512895 Triple 384-well, Single 96-Well Capacity Each
15532895 Quad 384-well Capacity Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 15569164 Supplier PCRmax Supplier No. 9394510

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.

PCRmax Alpha Cycler 4 Thermal Cycler has four independently controlled blocks for simplified temperature and space optimization

  • These compact thermal cyclers deliver reproducible results every run - with little to no training needed
  • Features include a clear responsive touch screen, secure user-specific programming, adjustable heated lid, and active sample cooling for sharper amplification
  • A Program Wizard generates a sequence specific protocol in seconds - a quick way to optimize new assays
  • Units also retain approximately 1000 reports for reviewing at a later date and run on an intuitive, HD Android™ tablet interface
  • Save personal settings to a USB drive; sign in is as simple as inserting your USB drive, no password needed
  • The four-block systems take up less space than two or four single-block units and cost less than purchasing multiple units
  • Control each block separately for added versatility and use by more than one person at a time, or run on the same program to increase throughput
  • Each block also has its own independent heated lid - no interruption of assays running at different settings

Specifications

Thermal Accuracy 50°C: ±0.25°C
Temperature Range (Metric) 10 to 100°C
Dimensions 33 x 70 x 53.5 cm
Capacity Single 384-well, Triple 96-Well Capacity
Electrical Requirements 100 to 240 VAC
Product Type Thermal Cycler
User Interface HD Android™ Tablet
Gradient Range 1° to 29°C
Heated Lid Yes, 35° to 115°C
Memory ≤ 1000 Programs
Block Configurations 4 Blocks
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.