Learn More
3M™ Aircare™ Filtration Unit ACU-03
Product Code. 15730058
Change view
Click to view available options
:
Dimensions (L x W x H): 0.333 x 0.233 x 0.18 m
Product Code.
15730058 Dimensions (L x W x H): 0.333 x 0.233 x 0.18 m
1 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 15730058 Supplier 3M™ Supplier No. 7100035217
Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
3M™ Aircare™ Filtration Unit ACU-03 provides high quality air from an external source of compressed air by removing water and oil droplets, solid particles, and odors.
  • Supplies the highest quality air to the Supplied Air Respirator System and compressed air tools by removing water and oil aerosols, particles and odors from the facility supplied compressed air
  • Wall mounted unit
  • Can be used for up to three users or two users plus air tool
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE Approved
Color Gray
Model ACU-03
Dimensions (L x W x H) 0.333 x 0.233 x 0.18 m
Product Type Wall-mounted Aircare Filtration Unit