Learn More
Brady™ Aggressive Adhesive Multipurpose Polyester Labels
Easily label components with the thermal transfer, glossy permanent polyester label material (B-422). Print almost anywhere with a durable Brady™ handheld label printer.
Brand:  Brady™ 170732
 View more versions of this product
Product Code. 17990680
  • € 54.55 / Each
Only null left
In Stock
Add to basket
This item is not returnable. View return policy
Description
Description
  • Designed for a wide range of applications such as labeling equipment, circuit boards, panel components and barcodes
  • Constructed to withstand abrasions, extreme temperatures and solvents such as alcohol, oil and water
  • Made with a permanent acrylic adhesive that sticks to rough, smooth and powder-coated flat surfaces
Specifications
Specifications
UL Recognised;CSA Accepted Compliant with Directive 2011/65/EU and 2015/863
White
Continuous
Label
Asset tracking;Barcode labelling;Component and equipment labelling;Data and telecommunications labelling;General identification;Inventory and inspection labelling
1
-40°C to 100°C
0.102 mm
25 ft.
0.5 in
M4C-500-422
Permanent Acrylic
Gloss
Polyester
Non-sterile
BMP41;BMP51;BMP53;M511;M410;M510
-40°F to 212°F
0.004 in
12.70 mm (W) x 7.62 m (L)
7.62 m
12.70 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.