missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agarose (Intermediate Melting/PCR Grade), Fisher BioReagents
For separation of low MW DNA ≤1000 bp by PCR analysis
Brand: Fisher BioReagents BP2410-100
2225.42 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Quantity | 100 g |
---|
This item is not returnable.
View return policy
Description

Specifications
Powder/Solid | |
Cream | |
DNase free | |
35.5°C max. | |
Poly Bottle | |
306.12 | |
Agarose, Intermediate Melting |
9012-36-6 | |
100 g | |
0.12 max. | |
1200g/cm2min. | |
C12H18O9 | |
DNase- and RNase-Free |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction