missing translation for 'onlineSavingsMsg'
Learn More

Color Coded High Resolution pH Buffer, Reagecon™
BIOPHARMA

Product Code. 11768465 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
500 mL
pH:
4
4.00
7
7.00
10.00
Unit Size:
500mL
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity pH unitSize
11768465 500 mL 7.00 500mL
11778465 500 mL 10.00 500mL
11494599 500 mL 4 500mL
11758465 500 mL 4.00 500mL
15214643 500 mL 7 Each
11404609 500 mL 7 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11768465 Supplier Reagecon Supplier No. 107000C

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Use color coded pH buffers for ease of use. Reagecon™ Color Coded High Resolution pH Buffers are tested at either 20°C or 25°C and are certified and NIST traceable.

  • Colored pH Buffer Solutions
  • High Resolution
  • Tested at 20°C or 25°C 
  • Certified by Reagecon’s ISO/IEC 17025 AccreditedTest Method
  • NIST traceable
  • Presented in various pack sizes

Specifications

Melting Point 0°C
Color Yellow
pH 7.00
Physical Form Liquid
Quantity 500 mL
Odor Characteristic
Solubility Information Soluble in water
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.