missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Techne™ Additional Loading Dock
Description
- This ergonomic plate loading dock allows for easy loading of a plate
- LED Backlight allows the user to easily visualise sample loading
- Plate adapter fits standard centrifuge rotors
Specifications
Specifications
| Content And Storage | 2 Centrifuge Adapters |
| For Use With (Equipment) | Prime Pro 48 Real-Time PCR System |
| Product Type | Additional Loading Dock |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?