Learn More
Cytiva AcroPak™ 200 Capsules with Supor™ Membrane
Product Code. 17194241
Click to view available options
Outlet Style:
1/4 in. MNPT Inlet, Stepped Hose Barb Outlet
Stepped Hose Barb
Unit Size:
Pack of 3
This item is not returnable. View return policy
This item is not returnable. View return policy
AcroPak capsules designed for research applications only.
  • Efficient processing of liquid volumes up to 20 L
  • Built-in prefilter extends filter life when particulate-laden solutions, such as serum-containing media, are processed
  • For buffers, tissue culture media, and other biological fluids
  • Newer technology recommended for simplified scale-up
  • High filtration area and compact size are ideal for upscale trials
TRUSTED_SUSTAINABILITY
Specifications
Material (Membrane) Hydrophilic PES
Pore Size 0.8/0.2 μm
Sterility Sterile
Filtration Area 200 cm2
Outlet Style Stepped Hose Barb
Quantity 3/Pk.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.