missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid, Glacial, HPLC Grade, J.T. Baker™
Shop All J.T.Baker Products
Click to view available options
Quantity:
2.5 L
Packaging:
Clear Glass Bottle
Unit Size:
2.5 L
Description
- For Use in High Performance Liquid Chromatography, Bulk Pharmaceutical Chemical
Specifications
Specifications
| CAS | 64-19-7 |
| Molecular Formula | C2H4O2 |
| MDL Number | MFCD00036152 |
| Synonym | ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure |
| InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
| SMILES | CC(O)=O |
| IUPAC Name | acetic acid |
| Molecular Weight (g/mol) | 60.05 |
| PubChem CID | 176 |
| ChEBI | CHEBI:15366 |
| Show More |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 4
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 3
- H228-Flammable solid.
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P370+P378-In case of fire: Use to extinguish.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction