missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid-d{4}, 99.5% (Isotopic)

Product Code. 11367928
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 1186-52-3 | C2H4O2 | 64.08 g/mol

Acetic acid-d{4} is used as a deuterated, polar and protic solvent, in food preservatives, PH control agent, flavor enhancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1186-52-3
Molecular Formula C2H4O2
Molecular Weight (g/mol) 64.08
MDL Number MFCD00051051
InChI Key QTBSBXVTEAMEQO-GUEYOVJQSA-N
Synonym acetic acid-d4, acetic-d3 acid-d, 2h3 acetic 2h acid, tetradeuteroacetic acid, 2 h? ethan 2 h oic acid, acetic acid d4, 2h4 acetic acid, cd3cood, acetic acid d, acetic acid-d4, 99.5 atom % d
PubChem CID 2723903
SMILES [2H]OC(=O)C([2H])([2H])[2H]

Specifications

Physical Form Liquid
Density 1.12 g/mL
Flash Point 40°C (104°F)
Odor Pungent, makes eyes water
Sensitivity Moisture
Quantity 10 g
Melting Point 17°C
Boiling Point 118°C
UN Number UN2789
Solubility Information Soluble in water,ethanol and ether.
IUPAC Name (²H₃)ethan(²H)oic acid
Formula Weight 64.08
Percent Purity 99.5%
Chemical Name or Material Acetic acid-d4, Isotopic
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.