Learn More
700 mOsm/kg Osmolality Standard, Reagecon
BIOPHARMA
Used to calibrate, control or validate osmometers
Brand:  Reagecon RE-OSM-700
Product Code. 16368185
  • 125.00 EUR / Pack of 12
Please to purchase this item. Need a web account? Register with us today!
Edge
This item is not returnable. View return policy
Description
Description
  • Manufactured in compliance with European, United States and Chinese Pharmacopoeia guidelines
  • Tested by ISO 17025 accredited test method
  • ISO 17025 accreditation for calibration of laboratory balances
  • Prepared gravimetrically on weight/weight basis
Specifications
Specifications
Amber glass ampoule
Calibration, Control or Validate Osmometer
Osmolality Standard
Liquid
Osmometer
12 x 5 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.