missing translation for 'onlineSavingsMsg'
Learn More

6-Bromohexanenitrile, 97%, Thermo Scientific Chemicals

Product Code. 11324649
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11324649 10 g 10g
11468583 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11324649 Supplier Thermo Scientific Alfa Aesar Supplier No. A10400.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2,6-Lutidine is used as a solvent in organic synthesis and as a sterically hindered mild base. It is also used as a vulcanization accelerator for dyes, resins and rubber. It also acts as a food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6621-59-6
Molecular Formula C6H10BrN
Molecular Weight (g/mol) 176.06
MDL Number MFCD00013834
InChI Key PHOSWLARCIBBJZ-UHFFFAOYSA-N
Synonym 6-bromocapronitrile, 6-bromohexanonitrile, hexanenitrile, 6-bromo, 5-bromopentyl cyanide, 6-bromo-hexanenitrile, 6-bromanylhexanenitrile, pubchem12505, acmc-1b5au, 6-bromohexanenitrile, phoswlarcibbjz-uhfffaoysa
PubChem CID 81093
IUPAC Name 6-bromohexanenitrile
SMILES BrCCCCCC#N

Specifications

Density 1.328
Boiling Point 134°C to 135°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4775
Quantity 10 g
UN Number UN3276
Beilstein 1744172
Formula Weight 176.06
Percent Purity 97%
Chemical Name or Material 6-Bromohexanenitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.