Learn More
Thermo Scientific™ 3M™ Comply™ Indicator Tape
Enables CSSD and OR personnel to know at a glance that the packs have been exposed to the sterilisation process. 3M™ Comply™ Indicator tape changes colour or turns darker when exposed to the sterilisation process.
€ 8.67 - € 25.90
Specifications
Certifications/Compliance | Meets NF EN 867-1 Class A, ISO 11140-1 Class 1 |
---|---|
Color | Beige |
For Use With (Application) | Sterilization verification |
For Use With (Equipment) | Autoclave |
Length (Metric) | 55 m |
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
10514885
|
Thermo Scientific™
7000136721 |
55 m x 19 mm | |||||||
11761351
|
Thermo Scientific™
7000138230 |
55 m x 24 mm |
€ 25.90
Each |
In Stock
Log in to see stock. ![]() |
|||||
12986976
|
Thermo Scientific™
7000138229 |
55 m x 18 mm |
€ 9.38
Each |
In Stock
Log in to see stock. ![]() |
|||||
11741605
|
Thermo Scientific™
1322V24 |
55 m x 24 mm | |||||||
Description
Enables CSSD and OR personnel to know at a glance that the packs have been exposed to the sterilisation process. 3M™ Comply™ Indicator tape changes colour or turns darker when exposed to the sterilisation process.
As an exposure monitor, 3M™ indicator tapes securely seal packs and enables CSSD and OR personnel to know at a glance that the packs have been exposed to the sterilisation process. The chemical indicator ink, printed in a diagonal striped pattern, changes colour or turns darker when exposed to the sterilisation process.- Lead-free steam indicator tape is designed for use on all wraps
- Reliable achesive adheres to packaging material
- Sustainable, solvent-free manufacturing proces
- Does not require hazardous waste disposal
- Redesigned packaging uses up to 30% less plastic
- Convenient, individually wrapped, easy-to-open rolls
- Lead free and no added latex
Specifications
Meets NF EN 867-1 Class A, ISO 11140-1 Class 1 | |
Sterilization verification | |
55 m | |
Lead-free, aggressive, pressure sensitive adhesive | |
Steam |
Beige | |
Autoclave | |
Indicator Tape | |
Yes |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.