missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Propanol, CHROMASOLV™ LC-MS, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
Unit Size:
1 L
2.5 L
Pack of 4
Pack of 6
Description
- For LC-MS and LC-UV trace metals analysis
- HPLC / UV-VIS quality control
- Low gradient baseline drift
- Low levels of trace metals
- No non-volatile impurities
Specifications
Specifications
| Color | Colorless |
| Density | 0.785g/mL at 25°C |
| Quantity | 1 L |
| Explosion Limit | 0.02 |
| Flash Point | 12°C (53.6°F) |
| Ignition Point | 750°F |
| Linear Formula | (CH3)2CHOH |
| Vapor Pressure | 44 mmHg (25°C), 33 mmHg (20°C) |
| UN Number | UN1219 |
| Beilstein | 635639 |
| Show More |
Safety and Handling
Product Identifier
- 2-Propanol
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : propan-2-ol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction