missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxyethanol, Reagent Grade, 99%, Honeywell™

Product Code. 15611200 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
25 L
Unit Size:
25 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15611200 25 L 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15611200 Supplier Honeywell Research Chemicals Supplier No. 12808225L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 110-80-5
Density 0.93g/mL at 25°C
Explosion Limit 0.14
Flash Point 42°C (107.6°F)
Ignition Point 460°F
Molecular Formula C4H10O2
Linear Formula C2H5OCH2CH2OH
MDL Number MFCD00002869
Quantity 25 L
UN Number UN1171
Beilstein 1098271
Synonym cellosolve, oxitol, ethyl cellosolve, ethylene glycol monoethyl ether, ethanol, 2-ethoxy, ethyl glycol, emkanol, hydroxy ether, ethylene glycol ethyl ether, dowanol ee
InChI Key ZNQVEEAIQZEUHB-UHFFFAOYSA-N
SMILES CCOCCO
IUPAC Name 2-ethoxyethanol
Molecular Weight (g/mol) 90.12
PubChem CID 8076
Vapor Density 3.1 (vs air)
ChEBI CHEBI:46788
Formula Weight 90.12g/mol
Vapor Pressure 3.8mmHg (20°C)
Percent Purity 99%
Grade ReagentPlus
Chemical Name or Material 2-Ethoxyethanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.