missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethanol-d{3}, 99% (Isotopic)

Product Code. 11386079
Click to view available options
Quantity:
1 g
5 x 1 g
Unit Size:
1g
5g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

NMR Solvent | CAS: 77253-67-9 | C2H3F3O | 103.06 g/mol

NMR Solvent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77253-67-9
Molecular Formula C2H3F3O
Molecular Weight (g/mol) 103.06
MDL Number MFCD00037682
InChI Key RHQDFWAXVIIEBN-IDPMSXFZSA-N
Synonym 2,2,2-trifluoroethanol-d3, 1,1-2h2-2,2,2-trifluoroetane-1-2h ol, ethan-1,1-d2-ol-d, 2,2,2-trifluoro, trifluoro 2 h? ethan 2 h ol, 2,2,2-trifluoroethanol-d3 >98 atom % d, 2,2,2-trifluoroethanol-d3, ≥99.5 atom % d, 2,2,2-trifluoroethanol-d3, 99 atom % d cp
PubChem CID 2724851
SMILES [2H]OC([2H])([2H])C(F)(F)F

Specifications

Physical Form Liquid
Density 1.45 g/mL
Packaging Prescored Ampule
Sensitivity Hygroscopic
Quantity 5 x 1 g
Melting Point -44°C
Boiling Point 77°C to 80°C
UN Number UN1992
IUPAC Name 1,1-dideuterio-1-deuteriooxy-2,2,2-trifluoroethane
Formula Weight 103.06
Percent Purity 99%
Chemical Name or Material 2,2,2-Trifluoroethanol-d3, Isotopic
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.