missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences 10x Buffer BXT

IBA Lifesciences GmbH 10x Buffer BXT; Strep-Tactin™XT Elution B. is a ready-to-use buffer for elution of Strep-tagged proteins from Strep-Tactin™ XT.

Brand:  IBA Lifesciences 2-1042-025


Product Code. 15878907

  • 1220.00 DKK / 25mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

  • 10x Buffer BXT is a 10x concentrated Strep-Tactin™XT elution buffer containing biotin.
  • Biotin is a water-soluble B-complex vitamin, also known as vitamin B7 or vitamin H.
  • Biotin binds to Strep-Tactin™XT with very high affinity but reversibly and thereby allows the efficient elution of Strep-tag™II or Twin-Strep-tag™ fusion proteins from the resin.

Please note that 10x Buffer BXT is not applicable for elution of Strep-tag™II or Twin-Strep-tag™ fusion proteins from Strep-Tactin™, since biotin irreversibly binds to these resins, and thus Strep-Tactin™ cannot be regenerated.

Specifications

Specifications

25 mL
25 mL
Biotin
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Buffer BXT (10x), Strep-Tactin®XT Biotin Elution Buffer

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Tris, EDTA

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.