missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dioxane-d{8}, 99% (Isotopic)

Product Code. 15421457
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 17647-74-4 | C4H8O2 | 96.155 g/mol

1,4-Dioxane-d{8} is used as a solvent in nuclear magnetic resonance (NMR) spectroscopy and acts as a stabilizer for 1,1,1-trichloroethane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Physical Form Liquid
Density 1.13 g/mL
Sensitivity Hygroscopic
Quantity 1 g
CAS 17647-74-4
Melting Point 12°C
Boiling Point 101°C
Molecular Formula C4H8O2
MDL Number MFCD00044239
UN Number UN1165
Synonym 1,4-dioxane-d8, 2h8-1,4-dioxane, 1,4-dioxane-d8, 99 atom % d, 2 h?-1,4-dioxane, 1,4-dioxane-2lh8, 1,4-dioxane d8, 1,4-dioxane-d8, ≥99 atom % d, 1,4-dioxane-d isotopic 1g, 1,4-dioxane-d8 >99.0 atom % d, 1 4-dioxane-d8 99 atom % d
Solubility Information Miscible with water.
InChI Key RYHBNJHYFVUHQT-SVYQBANQSA-N
SMILES C1COCCO1
IUPAC Name 2,2,3,3,5,5,6,6-octadeuterio-1,4-dioxane
Molecular Weight (g/mol) 96.155
PubChem CID 87209
Formula Weight 96.16
Percent Purity 99%
Chemical Name or Material 1,4-Dioxane-d8, Isotopic
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.