Product Code. 11379967
Click to view available options
Quantity:
1 g
5 g
5 x 1 g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
CAS: 17060-07-0 | C2H4Cl2 | 102.978 g/mol
1,2-Dichloroethane-d4 is widely used as a solvent for NMR analysis, and in the synthesis of poly(ethylene-d4 oxide) as a chain termination reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
CAS
17060-07-0
Molecular Formula
C2H4Cl2
Molecular Weight (g/mol)
102.978
MDL Number
MFCD00037531
InChI Key
WSLDOOZREJYCGB-LNLMKGTHSA-N
Synonym
1,2-dichloroethane-d4, 1,2-dichloroethane d4, ethylene-d4 dichloride, dichloro 2 h? ethane, 1,2-dichloroethane-d4, 99 atom % d, 1,2-dichloroethane-d4 98.0 atom % d, 1,2-dichloroethane-d4, analytical standard, 1,2-dichloroethane d4 2000 ng/microl in methanol
PubChem CID
12197860
IUPAC Name
1,2-dichloro-1,1,2,2-tetradeuterioethane
SMILES
C(CCl)Cl
Specifications
Physical Form
Liquid
Density
1.307 g/mL
Flash Point
15°C (60°F)
Refractive Index
1.443
Quantity
5 g
Melting Point
-35°C
Boiling Point
83°C
UN Number
UN1184
Solubility Information
Partially miscible with water.
Formula Weight
102.99
Percent Purity
99%
Chemical Name or Material
1,2-Dichloroethane-d4, Isotopic
Show More
Show Less
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.