missing translation for 'onlineSavingsMsg'
Learn More

1,1,2,2-Tetrachloroethane-d2, for NMR, 99 atom % D

Product Code. 10705801 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
20 mL
Packaging:
Glass Bottle
Unit Size:
20mL
5mL
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 33685-54-0 | C2H2Cl4 | 169.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR

Chemical Identifiers

CAS 33685-54-0
Molecular Formula C2H2Cl4
Molecular Weight (g/mol) 169.85
MDL Number MFCD00037672
InChI Key QPFMBZIOSGYJDE-QDNHWIQGSA-N
Synonym 1,1,2,2-tetrachloroethane-d2, 1,2-dideutero-1,1,2,2-tetrachloroethane, tetrachloro 2 h? ethane, 1,1,2,2-tetrachloroethane-d2 tce-d2, ethane-d2, 1,1,2,2-tetrachloro, ethane-1,2-d2, 1,1,2,2-tetrachloro, 1,1,2,2-tetrachloro-1,2-2h2 ethane, de87d, 1,1,2,2-tetrachloroethane-d2 >99.50 atom % d, 1,1,2,2-tetrachloroethane-d2, ≥99.5 atom % d
PubChem CID 118531
IUPAC Name 1,1,2,2-tetrachloro-1,2-dideuterioethane
SMILES C(C(Cl)Cl)(Cl)Cl

Specifications

Physical Form Liquid
Color Colorless
Density 1.6100g/mL
Deuterium (D) 98.5% min.
Packaging Glass Bottle
Refractive Index 1.4924
Quantity 5 mL
Boiling Point 146.0°C
Linear Formula (Cl2CD)2
Beilstein 01, III, 164
Formula Weight 169.86
Grade NMR
Chemical Name or Material 1,1,2,2-Tetrachloroethane-d2, 99 atom % D
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,1,2,2-Tetrachloroethane-d2
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H330-Fatal if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.
  • H310-Fatal in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.