Wipers
Shop Your Brands
Everything about Wipers
General Purpose Wipers
General Purpose Wipers
General purpose wipers are versatile workhorses used in manufacturing, the automotive industry, the food processing industry, and more. They tackle everything from routine cleaning to spill absorption with ease, enhancing efficiency and maintaining cleanliness standards.
Save on General Purpose Wipers
Wiping the Slate Clean: A Pro's Guide to Choosing the Perfect Wipes
Wiping the Slate Clean: A Pro's Guide to Choosing the Perfect Wipes
Learn more about wiper materials and uses with this short guide on wipes.
Read more about wipes
Controlled Environment Wipers
Controlled Environment Wipers
Cleanroom wipers, designed for controlled environments like laboratories and healthcare, are essential in preventing contamination, ensuring the integrity of experiments, and safeguarding patients' well-being.
Save on Controlled Environment Wipers
WypAll™ Product Selector
WypAllâ„¢ Product Selector
Download this comprehensive guide for WypAllâ„¢ wipers and always pick the right product for your task or application.
Download the document
Controlled Environment Wipers Buying Guide
Find the controlled environment wipe that best fits your requirements
1. Do you need sterile or non-sterile wipes?
Sterile wipes are vital for critical environments such as pharmaceutical or surgical settings, where they ensure minimal contamination. Non-sterile wipes are more cost-effective and suit less sensitive tasks, but may not meet stringent requirements.
2. Are you looking for presaturated or dry wipers?
The choice between pre-wetted and dry wipes in cleanrooms hinges on tasks, contamination control, and convenience. Pre-wetted wipes offer consistency, save time, and suit precise applications, while dry wipes allow customization, cost savings, and reduced waste.
3. Which wiper material best suits your needs?
Polyester wipes are synthetic and strong, feature low particle release , and have broad chemical compatibility, fitting cleanrooms. Cotton wipes are soft and absorbent but shed fibers. Biodegradable Lyocell wipes combine absorbency, strength, and eco-friendliness. Microfiber wipes blend polyester and polyamide, excelling at particle capture. Polyblend wipes merge polyester and cellulose for absorbency and cleanliness. Polyamide wipes provide strength and abrasion resistance. Polypropylene wipes are chemically resistant and lightweight, suiting cleanrooms.
General Purpose Wipers Buying Guide
Easily pick the wipe that is best suited for your environment and application
1. In what kind of environment do you work?
The choice of general purpose wipes hinges on the environment of use, which can impact their effectiveness. Diverse sectors have unique needs: industry requires compatibility with materials; automotive needs oil-resistant wipes; food processing demands food-safe options; laboratories need chemical compatibility and contamination control; and healthcare mandates pathogen-killing and patient safety. To avoid inefficiencies and compliance issues, it is crucial to align wipes with specific industry demands.
2. What is your task or application?
Choosing a general purpose wipe hinges on the specific tasks to be performed, which can affect their performance and cost-effectiveness. Needs include spill absorption, compatibility with detergents, specialty cleaning, and routine use. Task-specific considerations are crucial for efficient cleaning and equipment protection.
Our Top Selling Wipers at the Best Prices
Complete Your Order
Complete your wipers order with these supplies.
  • GREAT DEALS
    Barrier Wipes
    Products used to clean, disinfect, and absorb that include an impervious layer of material to prevent leakage of fluids and chemicals onto hands, gloves, and work surfaces. Available as wipes, liners, or rolls.
    Buy Now
  • GREAT DEALS
    Wipe Dispensers
    Containers designed to store and dispense wipes and towels. Available in various sizes, shapes, holding capacities, and material compositions. May have a wall-mounting feature.
    Buy Now
  • GREAT DEALS
    Labware Detergents and Cleaners
    Cleaning detergents designed to remove a range of contaminants from laboratory surfaces and equipment made of glass, plastic, and metal. Available as liquid solutions and powders for use in manual washing, ultrasonic cleaners, and dishwashers.
    Buy Now
  • GREAT DEALS
    Laboratory Disinfectants
    Cleaning products used to disinfect and deodorize hard and non-porous surfaces; available as liquid solutions, sprays, pads, and wipes. Product ingredients include alcohols, ammonium, and hydrogen peroxide and are suitable for cleanroom applications.
    Buy Now
Frequently Asked Questions
How do I ensure that the wipers I choose are compatible with the cleaning chemicals I use?
To ensure wiper compatibility with your cleaning chemicals, review the manufacturer's guidelines for both the wipers and the chemicals. Check for any specific instructions or restrictions regarding which wipers can be used with the cleaning agents. Look for information on material compatibility, chemical resistance, and any potential adverse reactions. Additionally, consider conducting a small-scale compatibility test by applying the cleaning chemical to a sample of the chosen wiper material and observing any adverse effects, such as deterioration, discoloration, or chemical reaction. Always prioritize safety and effectiveness when selecting wipers and cleaning chemicals for your specific cleaning tasks.
How do I properly store wipers to maintain their effectiveness?
To maintain the effectiveness of wipers, proper storage is crucial. Store wipers in their original, tightly-sealed containers to prevent moisture, dust, or contaminants from compromising their quality. Keep containers in a cool, dry place away from direct sunlight, extreme temperatures, or humidity, as these can degrade the wipers. Ensure that containers are clearly labeled to identify the type of wiper and any relevant expiration dates. Dispensing wipers should be done with clean hands or gloves to prevent contamination. Regularly inspect storage areas for cleanliness and any signs of damage or compromised seals, replacing damaged containers promptly. Proper storage preserves wiper quality and extends their shelf life.
What are the best practices for disposing of used wipers, especially in cleanrooms or sensitive environments?
Proper disposal of used wipers, particularly in cleanrooms or sensitive environments, is critical to preventing contamination and to maintain hygiene. Firstly, follow cleanroom-specific disposal guidelines if applicable. Place used wipers in designated bins or containers, ensuring they are clearly labeled for waste separation. Seal used wipers in plastic bags or containers to contain potential contaminants and odors. Dispose of them according to local regulations, adhering to guidelines for hazardous waste, recyclables, or general waste. Regular waste collection schedules should be established to prevent the accumulation of used wipes. Document disposal activities to demonstrate compliance and ensure a clean and controlled environment.