Thermo Scientific SureSTART Sample Handling Solutions
Find a Thermo Scientific™ SureSTART™ vial or well plate for nearly every GC or HPLC/UHPLC instrument, application and budget.
Three distinct SureSTART performance levels — differentiated by instrument compatibility, performance, reproducibility and value — make selecting the right product simple, clear and easy.
Why Choose Thermo Scientific SureSTART?
- The confidence of knowing every SureSTART product has undergone stringent testing for the desired performance level
- Instrument compatibility for all add-on and integrated chromatography autosamplers
- Sample security and low compound adsorption that protect valuable samples
- Three distinct performance levels to meet your price and performance needs
- Extend your consumables budget: The more you buy, the more you save
Request the Find a SKU/Easy Access Wheel to make finding the right vial simple.
SureSTART Vials and Well Plates: Accurate Results Start Here
Learn more about the three distinct SureSTART performance levels differentiated by instrument compatibility, performance, reproducibility and value.
Resources
Shop By SureSTART Performance Level
![]() | ![]() | ![]() | |
|---|---|---|---|
| Shop Now | Explore Performance Level 1 | Explore Performance Level 2 | Explore Performance Level 3 |
| Performance Level Overviews | |||
| When to Use | Cost-optimised products suitable for all HPLC/UHPLC and GC instrument types and tested for everyday analyses | High-throughput products for high-quality GC and HPLC/UHPLC data with an uninterrupted workflow | Typically packed in cleanrooms to protect against environmental factors, reduce low compound adsorption effects at trace levels, and contain the lowest amounts/levels of metals, extractables, and leachables; helping ensure you get the sensitivity and selectivity from your most challenging analytes confidently and consistently |
| Market Example | Academic/food analysis | Food safety, clinical, environmental, pharma, QC, industrial, forensics | Pharma/biopharma, clinical, pharma QC, omics |
| Instrument Type | Verifying compound presence and quantification using core detector technologies such as UV/Vis, PDA, RI, ELSD, FLD, FID, NPD, ECD, TCD, or FPD | Routine analyses via standard single or triple quadrupole MS systems | High-resolution accurate mass (HRAM) technologies, such as Thermo Scientific™ Orbitrap™ mass spectrometer or time of flight (ToF) MS/ion mobility (IMS) |
| Certification Options | Autosampler approved — tested to deliver performance, sample security and reproducibility | Specification certification by physical parameter | Certificate by physical parameters:
|
| Thermo Scientific SureSTART Offering | Vials: Glass, plastic, inserts Caps: AVCS™ caps, snap Well plates: WebSeal™ plastic Accessories | Vials: Glass, inserts, headspace, silanized, EPA, specification certified Caps: AVCS™ caps, bonded, magnetic, snap Kits: EPA, scintillation, crimpers, de-crimpers Well plates: WebSeal™ certified plastic Accessories | Vials: SureStop™ vials, gold-grade, headspace, fixed insert, micro-spring Caps: AVCS™ caps Certified kits: LC/GC, MS CERT, MS CERT+; TOC Electronic crimpers, decrimpers and station Well plates: WebSeal™ Plate+ (glass-coated) Accessories |
Additional Thermo Scientific Products
Simplifying Selection
Choosing the right vial depends on several set criteria: technique (e.g., LC, GC, or HS), instrument manufacturer, and whether the compounds being detected are light-sensitive or not (do you require clear or amber glass?).
Request our Find a SKU/Easy Access wheel for simplified vial selection when you complete the form.
- Quick lookup tool helps you find the right vial for your autosampler type
- One side features recommended part numbers for LC instruments and, the other, GC instruments
Our Fisher Scientific representatives are here to help. Learn more about how you can make your budget go further, or let us be your guide to determine which combination is most suitable for your application requirements.
Contact Us
Complete the form to request our Find a SKU/Easy Access wheel for simplified selection.