Buerkle Disposable samplers for foodstuffs
Disposable samplers for foodstuffs, blue, detectable, sterilised Scoops, spatulas, spoons and scrapers.
- Disposable sampler for foodstuffs blue or Detectable disposable samplers blue
- Especially for the food, animal feed and pharmaceutical industries
- Complies with EU food requirements and FDA regulations
- Individually packaged and sterilised by gamma rays
- Production, assembly and packaging according to clean room class 7
Key features
Disposable samplers for foodstuff (scoops, spatulas, spoons and scraper)
The shade of blue that is used does not occur in a natural form in foodstuffs. Because of this, blue products or parts of them can be quickly found and easily identified in a visual check if they are ever mixed into production.
- Food scoop, blue available from 25 ml to 1.000 ml (10 pieces per pack)
- Spatula for foodstuffs, blue: insertion depth 150 mm (10 pieces per pack)
- Spooon for foodstuffs, blue: 2.5 ml or 10 ml (100 pieces per pack)
- Scraper for foodstuffs, blue: 200 x 80 mm (10 pieces per pack
- Can be used as part of HACCP/IFS foreign object management
- Especially for the food, animal feed and pharmaceutical industries
- PS, blue
- Individually packaged and sterilised by gamma rays
- Production, assembly and packaging according to clean room class 7
- Complies with EU food requirements and FDA regulations
Detectable disposable samplers (scoops, spatulas, spoons and scraper)
These scoops, spatulas, spoons and scrapers are not only coloured blue but also have a special additive in the material that makes them visible to metal detectors or X rays. These products can therefore be rejected with standard systems for checking for foreign objects, even though they are made of plastic.
- Detectable scoop, blue available from 25 ml to 1.000 ml (10 pieces per pack)
- Detectable spatula, blue: insertion depth 150 mm (10 pieces per pack)
- Detectable spooon, blue: 2.5 ml or 10 ml (100 pieces per pack)
- Detectable scraper, blue: 200 x 80 mm (10 pieces per pack)
- Can be detected by colour scanners, metal detectors or X-rays
- PS detectable, blue
- Individually packaged and sterilised by gamma rays
- Production, assembly and packaging according to clean room class 7
- Complies with EU food requirements and FDA regulations