Electrodes and Accessories
Mettler Toledo™ KCl 3M Electrode Filling Solution
Mettler Toledo™ KCl 3M Electrode Filling Solution
Equipped with a special cap to facilitate easy filling of electrodes. Mettler Toledo™ KCl 3M Electrode Filling Solution is an electrolyte solution that enables optimum performance of pH and ion-selective electrodes. This bridge electrolyte contains no silver ions than can contaminate samples.
Thermo Scientific™ Orion™ pH Buffer Bottles
Thermo Scientific™ Orion™ pH Buffer Bottles
Perform accurate, reproducible pH calibrations with these color-coded buffer bottles.
Thermo Scientific™ Orion Star A Series Compact Printer
Thermo Scientific™ Orion Star A Series Compact Printer
Orion Star A series compact ink ribbon printer 100-240V, includes printer, one ink ribbon, one paper roll, meter interface cable and power cords (110V US/Japan, 220V Euro, 240V UK/Singapore and China/Australia/New Zealand)
Orion™ All-in-One™ pH Buffer Kits
Orion™ All-in-One™ pH Buffer Kits
Work with ease using this color-coded pH buffer kit that includes three buffers, storage solution and a storage bottle.
Mettler Toledo™ Economy Series pH and pH/ATC Combination Electrodes
Mettler Toledo™ Economy Series pH and pH/ATC Combination Electrodes
Offer outstanding price/performance ratio. Mettler Toledo Economy Series pH and pH/ATC Combination Electrodes are cost-effective alternative for less-demanding requirements.
Thermo Scientific™ Orion™ pH Electrode Storage Solutions
Thermo Scientific™ Orion™ pH Electrode Storage Solutions
Restore proper electrode performance and prolong electrode life using these pH electrode storage solutions.
pH Electrode Storage Solution, Reagecon™
pH Electrode Storage Solution, Reagecon™
Designed to maintain proper functioning and accurate readings of pH and reference electrodes. pH Electrode Storage Solution, Reagecon™ is a 3M KCl solution that helps maintain proper working conditions after cleaning or when the electrode has not been in use for an extended time period.
Thermo Scientific™ Orion™ Triode™ 3-in-1 pH/ATC Probe
Thermo Scientific™ Orion™ Triode™ 3-in-1 pH/ATC Probe
Operate this highly versatile probe, which contains a low-maintenance, gel-filled electrode with built-in ATC, in either the field or a plant.
Thermo Scientific™ Orion™ pH Electrode Filling Solution
Thermo Scientific™ Orion™ pH Electrode Filling Solution
Use this pH electrode filling solution with pH electrodes and reference electrodes.
Thermo Scientific™ Orion™ ISE Calibration Standards
Thermo Scientific™ Orion™ ISE Calibration Standards
Take reproducible ISE measurements with Thermo Scientific™ Orion™ ISE Calibration Standards. For use with ion selective electrodes from ammonia to water hardness.
Metrohm 3 mol/L KCI Electrolyte Solution
Metrohm 3 mol/L KCI Electrolyte Solution
An excellent solution for electrode conditioning.
METTLER TOLEDO™ Micro pH Electrodes
METTLER TOLEDO™ Micro pH Electrodes
pH electrodes designed for small sample volumes or for measuring in small containers.
METTLER TOLEDO™ InLab™ Science pH Electrodes
METTLER TOLEDO™ InLab™ Science pH Electrodes
A pH electrode featuring a special glass body which is resistant to corrosive or organic samples, and is ideal for high temperatures and biological media. The high electrolyte outflow prevents contaminants from entering and clogging the junction, which would interfere with the reading.
Thermo Scientific™ Orion™ ROSS Ultra™ Glass Triode™ pH/ATC Combination Electrodes
Thermo Scientific™ Orion™ ROSS Ultra™ Glass Triode™ pH/ATC Combination Electrodes
Combine a rapid temperature response with stability and precision by using Orion ROSS Ultra Glass Triode pH/ATC Combination Electrodes.
Thermo Scientific™ Orion™ pH Buffer Individual Use Pouches
Thermo Scientific™ Orion™ pH Buffer Individual Use Pouches
Ensure fresh, ready-to-use standards for pH calibration with these pH Buffer individual use pouches.
Thermo Scientific™ Orion™ 2 Cell Conductivity Probes
Thermo Scientific™ Orion™ 2 Cell Conductivity Probes
Perform applications in either the lab or the field with these versatile 2-electrode conductivity cells, available in a variety of materials and measurement ranges.
Thermo Scientific™ Orion™ Universal Power Adapter for Star A Meters
Thermo Scientific™ Orion™ Universal Power Adapter for Star A Meters
Ensure a constant power supply to your meter with the Thermo Scientific™ Orion™ Star Series Universal Power Adapter for Orion™ Star A series and legacy Orion™ Star series meters
Thermo Scientific™ Orion™ Star Series RS232 Printer and Computer Cable Set
Thermo Scientific™ Orion™ Star Series RS232 Printer and Computer Cable Set
Connect your computer and printer to a Versa Star, Star A200 or Star A300 series meter using the Thermo Scientific™ Orion™ Star Series RS232 Printer and Computer Cable Set.
Conductivity Standards for METTLER TOLEDO Conductivity Probes, METTLER TOLEDO™
Conductivity Standards for METTLER TOLEDO Conductivity Probes, METTLER TOLEDO™
Recommended NIST traceable solutions for calibrating or verifying METTLER TOLEDO conductivity electrodes.
METTLER TOLEDO™ InLab™ Expert pH Electrodes
METTLER TOLEDO™ InLab™ Expert pH Electrodes
A combination pH electrode with PEEK plastic shaft for chemical and mechanical durability, and a fixed cable. Designed for use in a variety of applications.
Fisherbrand™ Electrode Holder
Fisherbrand™ Electrode Holder
Handles multiple pH and conductivity readings quickly with reduced electrode breakage. Fisherbrand Electrode Holder accepts all brands of standard electrodes; extends 21 in. and pivots 360°; designed for smooth movement and effortless operation.
Thermo Scientific™ Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents
Thermo Scientific™ Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents
Take more accurate and reproducible ISE measurements with Thermo Scientific™ Orion™ ISE Ionic Strength Adjustors (ISA) and Special Reagents.
METTLER TOLEDO™ Ion Selective Electrodes (ISE)
METTLER TOLEDO™ Ion Selective Electrodes (ISE)
A selection of ISE probes for detecting concentration of various free ions.
Mettler Toledo™ SevenGo™ perfectION Combined ISE Electrodes
Mettler Toledo™ SevenGo™ perfectION Combined ISE Electrodes
Get an “All-In-One Solution” with SevenGo™ perfectION Combined ISE Electrodes. The brilliant Click & Clear™ press mechanism allows for quick cleaning and ensures stable readings even in difficult samples.
  spinner